Ethyl thiazole-2-carboxylate 97%

Supplier: Ambeed
Warning

A222678-100MG A222678-1G A222678-250MG A222678-5G
76786-040EA 8.49 USD
76786-040 76786-042 76786-044 76786-046
Ethyl thiazole-2-carboxylate 97%
Ethyl thiazole-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇NO₂S
MW: 157.1900 g/mol
MDL Number: MFCD06204689
CAS Number: 14527-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR