(4-Acetamidophenyl)boronic acid pinacol ester 95%

Supplier: Ambeed
Warning

A221880-1G A221880-250MG A221880-5G A221880-25G
76786-014EA 12 USD
76786-014 76786-016 76786-018 77872-133
(4-Acetamidophenyl)boronic acid pinacol ester 95%
(4-Acetamidophenyl)boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₀BNO₃
MDL Number: MFCD02093722
CAS Number: 214360-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR