Triethyleneglycol divinyl ether 95%

Supplier: Ambeed
Warning

A221371-25G A221371-100G A221371-500G
76785-984EA 7 USD
76785-984 77872-128 77872-129
Triethyleneglycol divinyl ether 95%
Triethyleneglycol divinyl ether

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₄
MDL Number: MFCD00074891
CAS Number: 765-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR