4-Cyanotetrahydro-4H-pyran 98%

Supplier: Ambeed
Warning

Synonyms: Oxane-4-carbonitrile

A221098-100G A221098-10G A221098-1G A221098-25G A221098-500G A221098-5G
76785-968EA 151.71 USD
76785-968 76785-970 76785-972 76785-974 76785-976 76785-978
4-Cyanotetrahydro-4H-pyran 98%
4-Cyanotetrahydro-4H-pyran

Precaución: for Research Use Only

Formula: C₆H₉NO
MW: 111.1400 g/mol
MDL Number: MFCD08690357
CAS Number: 4295-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR