4'-Pentyl-4-biphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-(4-Pentylphenyl)benzeneboronic acid, (4'-Pentyl-[1,1'-biphenyl]-4-yl)boronic acid

A220794-1G A220794-250MG
76785-964EA 18.04 USD
76785-964 76785-966
4'-Pentyl-4-biphenylboronic acid 98%
4'-Pentyl-4-biphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₇H₂₁BO₂
MW: 268.16 g/mol
MDL Number: MFCD07644474
CAS Number: 121554-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR