6-Amino-5-(2,2-diethoxyethyl)pyrimidin-4-ol 97%

Supplier: Ambeed
Warning

A220762-100MG A220762-1G A220762-250MG
76785-958EA 47.75 USD
76785-958 76785-960 76785-962
6-Amino-5-(2,2-diethoxyethyl)pyrimidin-4-ol 97%
6-Amino-5-(2,2-diethoxyethyl)pyrimidin-4-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₇N₃O₃
MW: 227.26 g/mol
MDL Number: MFCD02094191
CAS Number: 7400-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR