2-Bromo-5-tosyl-5H-pyrrolo[2,3-b]pyrazine 98%
Supplier: Ambeed
|
Warning
|
A220185-100MG
A220185-1G
A220185-250MG
76785-944EA
6
USD
76785-944
76785-946
76785-948
2-Bromo-5-tosyl-5H-pyrrolo[2,3-b]pyrazine 98%
2-Bromo-5-tosyl-5H-pyrrolo[2,3-b]pyrazine
Precaución: for Research Use Only
Formula:
C₁₃H₁₀BrN₃O₂S |
MDL Number:
MFCD12964053 CAS Number: 1201186-54-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...