Theaflavin 98%

Supplier: Ambeed
Warning

Synonyms: 3,4,6-Trihydroxy-1,8-bis(3α,5,7-trihydroxy-2α-chromanyl)-5H-benzocyclohepten-5-one, 3,4,6-Trihydroxy-1,8-bis((2R,3R)-3,5,7-trihydroxychroman-2-yl)-5H-benzo[7]annulen-5-one, 3,4,5-Trihydroxy-1,8-bis[(2R,3R)-3,5,7-trihydroxy-2-chromanyl]-6-benzo[7]annulenone

A218957-10MG A218957-1MG A218957-2MG A218957-5MG
76785-902EA 150 USD
76785-902 76785-904 76785-906 76785-908
Theaflavin 98%
Theaflavin

Precaución: for Research Use Only

Formula: C₂₉H₂₄O₁₂
MDL Number: MFCD03427500
CAS Number: 4670-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR