(1R,2R)-2-(Benzylamino)cyclohexanol 95%

Supplier: Ambeed
Warning

A218556-10G A218556-25G A218556-5G
76785-896EA 47 USD
76785-896 76785-898 76785-900
(1R,2R)-2-(Benzylamino)cyclohexanol 95%
(1R,2R)-2-(Benzylamino)cyclohexanol

Precaución: for Research Use Only

Formula: C₁₃H₁₉NO
MDL Number: MFCD09996881
CAS Number: 141553-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR