Tetramethylrhodamine ethyl ester perchlorate (TMRE) 97%

Supplier: Ambeed
Warning

Synonyms: Tetramethylrhodamine Ethyl Ester Perchlorate, TMRE, 3,6-bis(Dimethylamino)-9-[2-(ethoxycarbonyl)phenyl]-xanthylium perchlorate

A218540-1MG A218540-5MG A218540-10MG
76785-892EA 21.22 USD
76785-892 76785-894 77600-764
Tetramethylrhodamine ethyl ester perchlorate (TMRE) 97%
Tetramethylrhodamine ethyl ester perchlorate (TMRE)

Precaución: for Research Use Only

Formula: C₂₆H₂₇ClN₂O₇
MW: 514.95 g/mol
MDL Number: MFCD00467973
CAS Number: 115532-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR