5-Methoxy-1-methyl-1H-indole 98%

Supplier: Ambeed
Warning

A217963-100MG A217963-1G A217963-250MG A217963-5G
76785-876EA 21.22 USD
76785-876 76785-878 76785-880 76785-882
5-Methoxy-1-methyl-1H-indole 98%
5-Methoxy-1-methyl-1H-indole

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO
MW: 161.2 g/mol
MDL Number: MFCD05863250
CAS Number: 2521-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR