Spiro[4.5]decane-7,9-dione 97%
Supplier: Ambeed
|
Warning
|
A216809-100MG
A216809-1G
A216809-250MG
A216809-5G
76785-826EA
56
USD
76785-826
76785-828
76785-830
76785-832
Spiro[4.5]decane-7,9-dione 97%
Spiro[4.5]decane-7,9-dione
Precaución: for Research Use Only
Formula:
C₁₀H₁₄O₂ |
MDL Number:
MFCD03844397 CAS Number: 82683-51-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...