tert-Butyl-N-(3-thienyl)carbamate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl thiophen-3-ylcarbamate, 3-Aminothiophene, N-BOC protected

A215822-100G A215822-10G A215822-1G A215822-250MG A215822-25G A215822-5G
76785-804EA 1151.08 USD
76785-804 76785-806 76785-808 76785-810 76785-812 76785-814
tert-Butyl-N-(3-thienyl)carbamate 97%
tert-Butyl-N-(3-thienyl)carbamate

Precaución: for Research Use Only

Formula: C₉H₁₃NO₂S
MW: 199.27 g/mol
MDL Number: MFCD01928808
CAS Number: 19228-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR