Voglibose 98%

Supplier: Ambeed
Warning

Synonyms: (1S,2S,3R,4S,5S)-5-((1,3-Dihydroxypropan-2-yl)amino)-1-(hydroxymethyl)cyclohexane-1,2,3,4-tetraol

A214654-100MG A214654-10MG A214654-1MG A214654-250MG A214654-50MG A214654-1G
76785-770EA 31 USD
76785-770 76785-772 76785-774 76785-776 76785-778 77872-098
Voglibose 98%
Voglibose

Precaución: for Research Use Only

Formula: C₁₀H₂₁NO₇
MW: 267.28 g/mol
MDL Number: MFCD00865496
CAS Number: 83480-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR