2,2-Dimethyl-2H-pyrido[3,2-b][1,4]oxazin-3(4H)-one 98%

Supplier: Ambeed
Warning

A214198-100G A214198-1G A214198-25G A214198-5G
76785-762EA 271.6 USD
76785-762 76785-764 76785-766 76785-768
2,2-Dimethyl-2H-pyrido[3,2-b][1,4]oxazin-3(4H)-one 98%
2,2-Dimethyl-2H-pyrido[3,2-b][1,4]oxazin-3(4H)-one

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O₂
MW: 178.1879 g/mol
MDL Number: MFCD10000810
CAS Number: 20348-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR