Tetrapropylammonium bromide 98%
Supplier: Ambeed
|
Warning
|
A212478-100G
A212478-500G
A212478-25G
A212478-1000G
76785-718EA
22
USD
76785-718
76785-720
77087-370
77872-083
Tetrapropylammonium bromide 98%
Tetrapropylammonium bromide
Precaución: for Research Use Only
Formula:
C₁₂H₂₈BrN |
MDL Number:
MFCD00011840 CAS Number: 1941-30-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...