1-(2,3-Dihydro-7-benzofuranyl)ethanone 95%

Supplier: Ambeed
Warning

A211765-100MG A211765-1G A211765-5G A211765-250MG
76785-698EA 21 USD
76785-698 76785-700 76785-702 77872-080
1-(2,3-Dihydro-7-benzofuranyl)ethanone 95%
1-(2,3-Dihydro-7-benzofuranyl)ethanone

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₂
MDL Number: MFCD11656624
CAS Number: 170730-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR