tert-Butyl-3-hydroxy-4-methylenepiperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: N-t-BOC-4-Methylene-3-Piperidinol, tert-Butyl 3-hydroxy-4-methylenepiperidine-1-carboxylate

A211512-100MG A211512-1G A211512-250MG A211512-5G
76785-678EA 32.89 USD
76785-678 76785-680 76785-682 76785-684
tert-Butyl-3-hydroxy-4-methylenepiperidine-1-carboxylate 95%
tert-Butyl-3-hydroxy-4-methylenepiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₃
MW: 213.2735 g/mol
MDL Number: MFCD15474939
CAS Number: 159635-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR