6-(4-Methylpiperazin-1-yl)pyrimidin-4-amine 95%

Supplier: Ambeed
Warning

A208901-100MG A208901-1G A208901-250MG
76785-626EA 33.95 USD
76785-626 76785-628 76785-630
6-(4-Methylpiperazin-1-yl)pyrimidin-4-amine 95%
6-(4-Methylpiperazin-1-yl)pyrimidin-4-amine

Precaución: for Research Use Only

Formula: C9H15N5
MW: 193.2500 g/mol
MDL Number: MFCD09999229
CAS Number: 96225-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR