Vanillin isobutyrate 98%

Supplier: Ambeed
Warning

Synonyms: 4-Formyl-2-methoxyphenyl isobutyrate, Isobutavan

A208188-25G A208188-100G
76785-622EA 20 USD
76785-622 77872-053
Vanillin isobutyrate 98%
Vanillin isobutyrate

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₄
MW: 222.24 g/mol
MDL Number: MFCD00169858
CAS Number: 20665-85-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR