2-Phenyl-2,3-dihydroquinolin-4(1H)-one 97%

Supplier: Ambeed
Warning

A208103-10G A208103-1G A208103-250MG
76785-616EA 983.46 USD
76785-616 76785-618 76785-620
2-Phenyl-2,3-dihydroquinolin-4(1H)-one 97%
2-Phenyl-2,3-dihydroquinolin-4(1H)-one

Precaución: for Research Use Only

Formula: C₁₅H₁₃NO
MW: 223.27 g/mol
MDL Number: MFCD00098918
CAS Number: 16619-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR