Ethyl-6-bromonicotinate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-bromo-5-pyridinecarboxylate, Ethyl 6-bromonicotinate, 6-Bromonicotinic acid ethyl ester

A208088-10G A208088-1G A208088-250MG A208088-25G A208088-5G
76785-606EA 214.31 USD
76785-606 76785-608 76785-610 76785-612 76785-614
Ethyl-6-bromonicotinate 97%
Ethyl-6-bromonicotinate

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
MW: 230.0586 g/mol
MDL Number: MFCD11111853
CAS Number: 132334-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR