2,4-Dichloro-6-methyl-7H-pyrrolo[2,3-d]pyrimidine 98%

Supplier: Ambeed
Warning

A208020-1G A208020-250MG A208020-5G
76785-600EA 95 USD
76785-600 76785-602 76785-604
2,4-Dichloro-6-methyl-7H-pyrrolo[2,3-d]pyrimidine 98%
2,4-Dichloro-6-methyl-7H-pyrrolo[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₅Cl₂N₃
MDL Number: MFCD13193622
CAS Number: 1192711-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR