tert-Butyl-4-hydroxy-3-(hydroxymethyl)-1H-indole-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-hydroxy-3-(hydroxymethyl)-1H-indole-1-carboxylate

A207821-1G A207821-25G
76785-588EA 51.99 USD
76785-588 76785-590
tert-Butyl-4-hydroxy-3-(hydroxymethyl)-1H-indole-1-carboxylate 95%
tert-Butyl-4-hydroxy-3-(hydroxymethyl)-1H-indole-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₇NO₄
MW: 263.2900 g/mol
MDL Number: MFCD05864722
CAS Number: 914349-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR