2-Benzyl-1H-benzimidazole 97%

Supplier: Ambeed
Warning

A207724-10G A207724-25G A207724-5G
76785-574EA 25 USD
76785-574 76785-576 76785-578
2-Benzyl-1H-benzimidazole 97%
2-Benzyl-1H-benzimidazole

Precaución: for Research Use Only

Formula: C₁₄H₁₂N₂
MDL Number: MFCD00022680
CAS Number: 621-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR