Methyl-3-amino-6-chloropyrazine-2-carboxylate 98%
Supplier: Ambeed
|
Warning
|
A207024-100MG
A207024-10G
A207024-1G
A207024-250MG
A207024-5G
76785-544EA
6
USD
76785-544
76785-546
76785-548
76785-550
76785-552
Methyl-3-amino-6-chloropyrazine-2-carboxylate 98%
Methyl-3-amino-6-chloropyrazine-2-carboxylate
Precaución: for Research Use Only
Formula:
C₆H₆ClN₃O₂ |
MDL Number:
MFCD00834965 CAS Number: 1458-03-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...