5-Bromo-[1,3]thiazolo[5,4-b]pyridin-2-amine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromothiazolo[5,4-b]pyridin-2-amine

A206658-1G A206658-5G
76785-526EA 74.27 USD
76785-526 76785-528
5-Bromo-[1,3]thiazolo[5,4-b]pyridin-2-amine 98%
5-Bromo-[1,3]thiazolo[5,4-b]pyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₄BrN₃S
MW: 230.09 g/mol
MDL Number: MFCD11846492
CAS Number: 934266-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR