7-Bromo-1-methyl-1H-quinoxalin-2-one 95%

Supplier: Ambeed
Warning

A206593-100MG A206593-250MG A206593-50MG A206593-1G
76785-520EA 34 USD
76785-520 76785-522 76785-524 77667-732
7-Bromo-1-methyl-1H-quinoxalin-2-one 95%
7-Bromo-1-methyl-1H-quinoxalin-2-one

Precaución: for Research Use Only

Formula: C₉H₇BrN₂O
MDL Number: MFCD10000855
CAS Number: 82019-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR