trans,trans-4-[(E)-1-Propenyl]-4'-propylbicyclohexyl 97%

Supplier: Ambeed
Warning

Synonyms: (trans,trans)-4-(Prop-1-en-1-yl)-4'-propyl-1,1'-bi(cyclohexane)

A206265-1G A206265-5G
76785-514EA 10 USD
76785-514 76785-516
trans,trans-4-[(E)-1-Propenyl]-4'-propylbicyclohexyl 97%
trans,trans-4-[(E)-1-Propenyl]-4'-propylbicyclohexyl

Precaución: for Research Use Only

Formula: C₁₈H₃₂
MW: 248.45 g/mol
MDL Number: MFCD12761279
CAS Number: 279246-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR