Z-D-Arg-OH·HCl 97%

Supplier: Ambeed
Warning

A205903-5G A205903-1G A205903-25G
76785-496EA 34 USD
76785-496 77667-722 77872-033
Z-D-Arg-OH·HCl 97%
Z-D-Arg-OH·HCl

Precaución: for Research Use Only

Formula: C₁₄H₂₁ClN₄O₄
MDL Number: MFCD00063010
CAS Number: 113712-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR