Tacrine hydrochloride 99%

Supplier: Ambeed
Warning

Synonyms: 1,2,3,4-Tetrahydroacridin-9-amine hydrochloride, 9-Amino-1,2,3,4-tetrahydroacridine hydrochloride, THA hydrochloride

A205226-1G A205226-1MG
76785-470EA 28.65 USD
76785-470 76785-472
Tacrine hydrochloride 99%
Tacrine hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₁₄N₂·HCl
MW: 234.72 g/mol
MDL Number: MFCD00012657
CAS Number: 1684-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR