5-Amino-6-fluoro-1H-indazole 96%

Supplier: Ambeed
Warning

A205213-100MG A205213-250MG
76785-462EA 80 USD
76785-462 76785-464 77872-026
5-Amino-6-fluoro-1H-indazole 96%
5-Amino-6-fluoro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₆FN₃
MDL Number: MFCD09261137
CAS Number: 709046-14-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR