tert-Butyl 4-bromo-2-methylbenzoate 98%

Supplier: Ambeed
Warning

A205142-100MG A205142-1G A205142-250MG A205142-5G
76785-452EA 40.32 USD
76785-452 76785-454 76785-456 76785-458
tert-Butyl 4-bromo-2-methylbenzoate 98%
tert-Butyl 4-bromo-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₅BrO₂
MW: 271.15 g/mol
MDL Number: MFCD08703427
CAS Number: 445003-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR