TGX-221 98+%

Supplier: Ambeed
Warning

Synonyms: 7-Methyl-2-morpholino-9-(1-(phenylamino)ethyl)-4H-pyrido[1,2-a]pyrimidin-4-one

A203716-100MG A203716-10MG A203716-1MG A203716-50MG A203716-5MG
76785-436EA 217 USD
76785-436 76785-438 76785-440 76785-442 76785-444
TGX-221 98+%
TGX-221

Precaución: for Research Use Only

Formula: C₂₁H₂₄N₄O₂
MDL Number: MFCD11113209
CAS Number: 663619-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR