Ethyl-2-amino-6-methylbenzoate 98+%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-amino-6-methylbenzoate

A203352-1G A203352-250MG A203352-5G
76785-426EA 215.37 USD
76785-426 76785-428 76785-430
Ethyl-2-amino-6-methylbenzoate 98+%
Ethyl-2-amino-6-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
MDL Number: MFCD14631083
CAS Number: 90259-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR