1,1,1-Trifluoropropan-2-amine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 1,1,1-Trifluoropropan-2-amine hydrochloride, 2-Amino-1,1,1-trifluoropropane hydrochloride

A202931-250MG A202931-25G A202931-5G A202931-1G
76785-408EA 15.92 USD
76785-408 76785-410 76785-412 77567-932
1,1,1-Trifluoropropan-2-amine hydrochloride 98%
1,1,1-Trifluoropropan-2-amine hydrochloride

Precaución: For research use only.

Formula: C₃H₆F₃N·ClH
MW: 149.543 g/mol
MDL Number: MFCD04972986
CAS Number: 2968-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR