1-Chloro-2,4-difluoro-3-nitrobenzene 97%

Supplier: Ambeed
Warning

A202521-100MG A202521-10G A202521-1G A202521-250MG A202521-5G
76785-386EA 41.38 USD
76785-386 76785-388 76785-390 76785-392 76785-394
1-Chloro-2,4-difluoro-3-nitrobenzene 97%
1-Chloro-2,4-difluoro-3-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂ClF₂NO₂
MW: 193.54 g/mol
MDL Number: MFCD10686546
CAS Number: 1151767-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR