Methyl-5-chloro-1H-pyrrolo[2,3-b]pyridine-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-chloro-1H-pyrrolo[2,3-b]pyridine-2-carboxylate

A202242-100MG A202242-1G A202242-250MG
76785-376EA 32.89 USD
76785-376 76785-378 76785-380
Methyl-5-chloro-1H-pyrrolo[2,3-b]pyridine-2-carboxylate 98%
Methyl-5-chloro-1H-pyrrolo[2,3-b]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇ClN₂O₂
MW: 210.6200 g/mol
MDL Number: MFCD09841660
CAS Number: 952182-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR