1,5-Dimethyl-1H-pyrazol-4-amine 95%

Supplier: Ambeed
Warning

A202149-5G A202149-250MG
76785-374EA 240.83 USD
76785-374 77667-682
1,5-Dimethyl-1H-pyrazol-4-amine 95%
1,5-Dimethyl-1H-pyrazol-4-amine

Precaución: for Research Use Only

Formula: C₅H₉N₃
MW: 111.14 g/mol
MDL Number: MFCD02253774
CAS Number: 121983-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR