2-Cyano-4-methyl biphenyl 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methyl-[1,1'-biphenyl]-2-carbonitrile

A201568-10G A201568-25G A201568-5G
76785-360EA 6.37 USD
76785-360 76785-362 76785-364
2-Cyano-4-methyl biphenyl 98%
2-Cyano-4-methyl biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₁N
MW: 193.244 g/mol
MDL Number: MFCD06411093
CAS Number: 64113-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR