trans-4-(Boc-amino)cyclohexaneacetaldehyde 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-butyl trans-N-[4-(2-oxoethyl)cyclohexyl]carbamate, tert-Butyl (trans-4-(2-oxoethyl)cyclohexyl)carbamate
A201451-1G
A201451-250MG
A201451-5G
A201451-100MG
76785-354EA
104
USD
76785-354
77600-642
77667-672
77872-006
trans-4-(Boc-amino)cyclohexaneacetaldehyde 98%
trans-4-(Boc-amino)cyclohexaneacetaldehyde
Precaución: for Research Use Only
Formula:
C₁₃H₂₃NO₃ MW: 241.331 g/mol |
MDL Number:
MFCD03844592 CAS Number: 215790-29-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...