Fmoc-D-4-pyridylalanine 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-D-4-Pal-OH

A201254-100MG A201254-10G A201254-1G A201254-250MG A201254-25G A201254-5G
76785-338EA 7 USD
76785-338 76785-340 76785-342 76785-344 76785-346 76785-348
Fmoc-D-4-pyridylalanine 95%
Fmoc-D-4-pyridylalanine

Precaución: for Research Use Only

Formula: C₂₃H₂₀N₂O₄
MDL Number: MFCD00672567
CAS Number: 205528-30-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR