5,7-Dihydro-2-(methylsulfonyl)-6H-pyrrolo[3,4-d]pyrimidine, N6-BOC protected 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl 2-(methylsulphonyl)-5H-pyrrolo[3,4-d]pyrimidine-6(7H)-carboxylate, tert-Butyl 5,7-dihydro-2-(methylsulphonyl)-6H-pyrrolo[3,4-d]pyrimidine-6-carboxylate
A200583-5G
A200583-100MG
76785-324EA
1431.16
USD
76785-324
77667-666
5,7-Dihydro-2-(methylsulfonyl)-6H-pyrrolo[3,4-d]pyrimidine, N6-BOC protected 97%
5,7-Dihydro-2-(methylsulfonyl)-6H-pyrrolo[3,4-d]pyrimidine, N6-BOC protected
Precaución: for Research Use Only
Formula:
C₁₂H₁₇N₃O₄S MW: 299.346 g/mol |
MDL Number:
MFCD09839781 CAS Number: 365996-87-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...