4-Methoxybenzophenone 98%

Supplier: Ambeed
Warning

Synonyms: p-Methoxybenzophenone, (4-Methoxyphenyl)(phenyl)methanone

A200487-1000G A200487-100G A200487-10G A200487-500G A200487-25G
76785-310EA 169.75 USD
76785-310 76785-312 76785-314 76785-316 77302-686
4-Methoxybenzophenone 98%
4-Methoxybenzophenone

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
MDL Number: MFCD00008403
CAS Number: 611-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR