4,4'-((1S,3aR,4S,6aR)-Hexahydrofuro[3,4-c]furan-1,4-diyl)bis(2-methoxyphenol) 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Pinoresinol
A200331-10MG
A200331-1MG
A200331-5MG
A200331-100MG
A200331-50MG
76785-304EA
88.06
USD
76785-304
76785-306
76785-308
77667-658
77667-660
4,4'-((1S,3aR,4S,6aR)-Hexahydrofuro[3,4-c]furan-1,4-diyl)bis(2-methoxyphenol) 95%
4,4'-((1S,3aR,4S,6aR)-Hexahydrofuro[3,4-c]furan-1,4-diyl)bis(2-methoxyphenol)
Precaución: for Research Use Only
Formula:
C₂₀H₂₂O₆ MW: 358.38 g/mol |
MDL Number:
MFCD07784516 CAS Number: 487-36-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...