Methyl-5-bromothiazole-4-carboxylate 98%

Supplier: Ambeed
Warning

A199963-10G A199963-1G A199963-250MG A199963-25G A199963-5G
76785-286EA 56 USD
76785-286 76785-288 76785-290 76785-292 76785-294
Methyl-5-bromothiazole-4-carboxylate 98%
Methyl-5-bromothiazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₅H₄BrNO₂S
MDL Number: MFCD08275713
CAS Number: 913836-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR