Methyl-2-amino-5-methylbenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-amino-5-methylbenzoate

A199903-100G A199903-10G A199903-1G A199903-25G A199903-5G
76785-276EA 95.49 USD
76785-276 76785-278 76785-280 76785-282 76785-284
Methyl-2-amino-5-methylbenzoate 97%
Methyl-2-amino-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MW: 165.1900 g/mol
MDL Number: MFCD08234606
CAS Number: 18595-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR