N-n-Octyl-D-glucamine 97%

Supplier: Ambeed
Warning

Synonyms: (2R,3R,4R,5S)-6-(Octylamino)hexane-1,2,3,4,5-pentaol, 1-Deoxy-1-(n-octylamino)-D-glucitol

A199820-1000G A199820-100G A199820-25G A199820-500G A199820-5G
76785-266EA 87 USD
76785-266 76785-268 76785-270 76785-272 76785-274
N-n-Octyl-D-glucamine 97%
N-n-Octyl-D-glucamine

Precaución: for Research Use Only

Formula: C₁₄H₃₁NO₅
MW: 293.40 g/mol
MDL Number: MFCD00134352
CAS Number: 23323-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR