(S)-2-((2-Amino-6-oxo-1H-purin-9(6H)-yl)methoxy)ethyl 2-amino-3-methylbutanoate hydrochloride 97%

Supplier: Ambeed
Warning

A199102-10MG A199102-1G A199102-1MG A199102-25G A199102-50MG A199102-5G A199102-250MG
76785-236EA 9 USD
76785-236 76785-238 76785-240 76785-242 76785-244 76785-246 77667-654
(S)-2-((2-Amino-6-oxo-1H-purin-9(6H)-yl)methoxy)ethyl 2-amino-3-methylbutanoate hydrochloride 97%
(S)-2-((2-Amino-6-oxo-1H-purin-9(6H)-yl)methoxy)ethyl 2-amino-3-methylbutanoate hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₂₁ClN₆O₄
MDL Number: MFCD11114398
CAS Number: 124832-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR