2-Amino-3,5-difluorobenzoic acid 98%

Supplier: Ambeed
Warning

A198662-10G A198662-1G A198662-250MG A198662-25G A198662-5G
76785-214EA 106.09 USD
76785-214 76785-216 76785-218 76785-220 76785-222
2-Amino-3,5-difluorobenzoic acid 98%
2-Amino-3,5-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅F₂NO₂
MW: 173.1200 g/mol
MDL Number: MFCD01569541
CAS Number: 126674-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR